Tisk…

5-Amino-2-isopropyl-2H-indazole

Dodavatel: Apollo Scientific

APOSOR470057-5GEA 31910 CZK
APOSOR470057-5G
5-Amino-2-isopropyl-2H-indazole
5-Amino-2-isopropyl-2H-indazole
Vzorec: C₁₀H₁₃N₃
MW: 175,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375232
CAS číslo: 1261268-91-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR