3-Bromo-4-piperazinobenzoic acid
Dodavatel: Apollo Scientific
APOSOR470063-5GEA
22920
CZK
APOSOR470063-5G
3-Bromo-4-piperazinobenzoic acid
3-Bromo-4-piperazinobenzoic acid
Vzorec:
C₁₁H₁₃BrN₂O₂ MW: 285,137 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11110935 CAS číslo: 1131594-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...