Tisk…

4-(Methylsulphonyl)-3-piperazinobenzoic acid

Dodavatel: Apollo Scientific
Varování

APOSOR470065-5GEA 33400 CZK
APOSOR470065-5G
4-(Methylsulphonyl)-3-piperazinobenzoic acid
4-(Methylsulphonyl)-3-piperazinobenzoic acid
Vzorec: C₁₂H₁₆N₂O₄S
MW: 284,331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13191609
CAS číslo: 1197193-03-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR