2-(Boc-amino)ethoxylbenzonitrile
Dodavatel: Apollo Scientific
APOSOR470128-5GEA
16830
CZK
APOSOR470128-5G
2-(Boc-amino)ethoxylbenzonitrile
2-(Boc-amino)ethoxylbenzonitrile
Vzorec:
C₁₄H₁₈N₂O₃ MW: 262,304 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18070983 CAS číslo: 263409-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...