2-(Methylsulphonamido)phenylacetonitrile
Dodavatel: Apollo Scientific
APOSOR470136-5GEA
17130
CZK
APOSOR470136-5G
2-(Methylsulphonamido)phenylacetonitrile
2-(Methylsulphonamido)phenylacetonitrile
Vzorec:
C₉H₁₀N₂O₂S MW: 210,253 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18070996 CAS číslo: 36268-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...