Tisk…

2-(Cyclopentyloxy)acetamide

Dodavatel: Apollo Scientific

APOSOR470140-5GEA 26090 CZK
APOSOR470140-5G
2-(Cyclopentyloxy)acetamide
2-(Cyclopentyloxy)acetamide
Vzorec: C₇H₁₃NO₂
MW: 143,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16042597
CAS číslo: 855929-18-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR