Tisk…

Sodium 3-Cyanopropane-1-sulphonate

Dodavatel: Apollo Scientific

APOSOR470147-5GEA 28700 CZK
APOSOR470147-5G
Sodium 3-Cyanopropane-1-sulphonate
Sodium 3-Cyanopropane-1-sulphonate
Vzorec: C₄H₆NO₃S·Na
MW: 171,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18071013
CAS číslo: 35880-67-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR