Methyl 3-chloro-4-(4-methylpiperazino)benzoate
Dodavatel: Apollo Scientific
APOSOR470163-5GEA
17130
CZK
APOSOR470163-5G
Methyl 3-chloro-4-(4-methylpiperazino)benzoate
Methyl 3-chloro-4-(4-methylpiperazino)benzoate
Vzorec:
C₁₃H₁₇ClN₂O₂ MW: 268,739 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18071042 CAS číslo: 1256633-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...