Tisk…

Methyl N-Boc-2-bromo-5-sulphamoylbenzoate

Dodavatel: Apollo Scientific

APOSOR470177-1GEA 11310 CZK
APOSOR470177-1G
Methyl N-Boc-2-bromo-5-sulphamoylbenzoate
Methyl N-Boc-2-bromo-5-sulphamoylbenzoate
Vzorec: C₁₃H₁₆BrNO₆S
MW: 394,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18651694
CAS číslo: 1272756-19-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR