Methyl 2-(5-Amino-4-cyano-2-furyl)benzoate
Dodavatel: Apollo Scientific
APOSOR470190-5GEA
28700
CZK
APOSOR470190-5G
Methyl 2-(5-Amino-4-cyano-2-furyl)benzoate
Methyl 2-(5-Amino-4-cyano-2-furyl)benzoate
Vzorec:
C₁₃H₁₀N₂O₃ MW: 242,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18375285 CAS číslo: 1261268-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...