Tisk…

6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one

Dodavatel: Apollo Scientific

APOSOR470227-5GEA 31910 CZK
APOSOR470227-5G
6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
Vzorec: C₁₂H₁₃BrN₂O
MW: 281,148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18651733
CAS číslo: 1272756-17-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR