Tisk…

6-Chloro-5-methyl-1H-indazole

Dodavatel: Apollo Scientific

APOSOR470242-1GEA 3540 CZK
APOSOR470242-1G
6-Chloro-5-methyl-1H-indazole
6-Chloro-5-methyl-1H-indazole
Vzorec: C₈H₇ClN₂
MW: 166,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09880179
CAS číslo: 1000341-02-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR