Tisk…

3-Iodo-1-methyl-4-nitro-1H-pyrrolo[2,3-b]pyridine

Dodavatel: Apollo Scientific

APOSOR470252-1GEA 43640 CZK
APOSOR470252-1G
3-Iodo-1-methyl-4-nitro-1H-pyrrolo[2,3-b]pyridine
3-Iodo-1-methyl-4-nitro-1H-pyrrolo[2,3-b]pyridine
Vzorec: C₈H₆IN₃O₂
MW: 303,057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22423101
CAS číslo: 1398534-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR