Ethyl 3-(1,4-Benzodioxan-2-yl)-3-oxopropanoate
Dodavatel: Apollo Scientific
APOSOR470283-5GEA
31910
CZK
APOSOR470283-5G
Ethyl 3-(1,4-Benzodioxan-2-yl)-3-oxopropanoate
Ethyl 3-(1,4-Benzodioxan-2-yl)-3-oxopropanoate
Vzorec:
C₁₃H₁₄O₅ MW: 250,247 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07370064 CAS číslo: 889955-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...