2-Amino-6-chloro-3-methylbenzoic acid
Dodavatel: Apollo Scientific
APOSOR470333-5GEA
31910
CZK
APOSOR470333-5G
2-Amino-6-chloro-3-methylbenzoic acid
2-Amino-6-chloro-3-methylbenzoic acid
Vzorec:
C₈H₈ClNO₂ MW: 185,608 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06409109 CAS číslo: 857005-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...