Tisk…

5-Methyl-2-(phenylethynyl)pyridine

Dodavatel: Apollo Scientific

APOSOR470351-1GEA 8960 CZK
APOSOR470351-1G
5-Methyl-2-(phenylethynyl)pyridine
5-Methyl-2-(phenylethynyl)pyridine
Vzorec: C₁₄H₁₁N
MW: 193,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18205120
CAS číslo: 124300-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR