Tisk…

2-Hydroxy-5-pentylbenzoic acid

Dodavatel: Apollo Scientific

APOSOR470353-5GEA 31910 CZK
APOSOR470353-5G
2-Hydroxy-5-pentylbenzoic acid
2-Hydroxy-5-pentylbenzoic acid
Vzorec: C₁₂H₁₆O₃
MW: 208,254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11203636
CAS číslo: 28488-46-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR