Tisk…

Ethyl 3-Oxo-4-(1-Boc-4-piperidyl)butyrate

Dodavatel: Apollo Scientific

APOSOR470357-5GEA 40010 CZK
APOSOR470357-5G
Ethyl 3-Oxo-4-(1-Boc-4-piperidyl)butyrate
Ethyl 3-Oxo-4-(1-Boc-4-piperidyl)butyrate
Vzorec: C₁₆H₂₇NO₅
MW: 313,389 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07370059
CAS číslo: 916791-39-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR