Tisk…

6-Phenyl-2-piperidinecarboxylic acid

Dodavatel: Apollo Scientific

APOSOR470374-1GEA 35050 CZK
APOSOR470374-1G
6-Phenyl-2-piperidinecarboxylic acid
6-Phenyl-2-piperidinecarboxylic acid
Vzorec: C₁₂H₁₅NO₂
MW: 205,253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410516
CAS číslo: 1219143-12-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR