5-(3,4-Dimethoxyphenyl)nicotinic acid
Dodavatel: Apollo Scientific
APOSOR470376-5GEA
55220
CZK
APOSOR470376-5G
5-(3,4-Dimethoxyphenyl)nicotinic acid
5-(3,4-Dimethoxyphenyl)nicotinic acid
Vzorec:
C₁₄H₁₃NO₄ MW: 259,257 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06410296 CAS číslo: 887973-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...