6-Cbz-2,2-dimethylhexahydropyrrolo[3,4-d]-1,3-oxazine
Dodavatel: Apollo Scientific
APOSOR470384-1GEA
25530
CZK
APOSOR470384-1G
6-Cbz-2,2-dimethylhexahydropyrrolo[3,4-d]-1,3-oxazine
6-Cbz-2,2-dimethylhexahydropyrrolo[3,4-d]-1,3-oxazine
Vzorec:
C₁₆H₂₂N₂O₃ MW: 290,357 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20134214 CAS číslo: 1346597-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...