Tisk…

(E)-3-(3-tert-Butoxyphenyl)acrylic acid

Dodavatel: Apollo Scientific

APOSOR470400-5GEA 37690 CZK
APOSOR470400-5G
(E)-3-(3-tert-Butoxyphenyl)acrylic acid
(E)-3-(3-tert-Butoxyphenyl)acrylic acid
Vzorec: C₁₃H₁₆O₃
MW: 220,264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412398
CAS číslo: 898405-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR