Tisk…

Tri-tert-butylmethanol

Dodavatel: Apollo Scientific

APOSOR470411-250MGEA 6320 CZK
APOSOR470411-250MG
Tri-tert-butylmethanol
Tri-tert-butylmethanol
Vzorec: C₁₃H₂₈O
MW: 200,361 g/mol
Bod tání: 94…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00204381
CAS číslo: 41902-42-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR