Tisk…

Ethyl 3-(2-Bromo-6-nitrophenyl)-2-oxopropanoate

Dodavatel: Apollo Scientific

APOSOR470416-5GEA 11310 CZK
APOSOR470416-5G
Ethyl 3-(2-Bromo-6-nitrophenyl)-2-oxopropanoate
Ethyl 3-(2-Bromo-6-nitrophenyl)-2-oxopropanoate
Vzorec: C₁₁H₁₀BrNO₅
MW: 316,105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20491407
CAS číslo: 608510-29-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR