Tisk…

(4-Biphenylylmethyl)hydrazine hydrochloride

Dodavatel: Apollo Scientific

APOSOR470424-1GEA 6880 CZK
APOSOR470424-1G
(4-Biphenylylmethyl)hydrazine hydrochloride
(4-Biphenylylmethyl)hydrazine hydrochloride
Vzorec: C₁₃H₁₄N₂·ClH
MW: 234,725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01722362
CAS číslo: 75333-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR