Tisk…

1-Allyl-7-methoxy-2-tetralone

Dodavatel: Apollo Scientific

APOSOR470455-5GEA 28700 CZK
APOSOR470455-5G
1-Allyl-7-methoxy-2-tetralone
1-Allyl-7-methoxy-2-tetralone
Vzorec: C₁₄H₁₆O₂
MW: 216,276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21364428
CAS číslo: 29093-46-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR