Tisk…

Methyl 1-Cbz-3-allylpiperidine-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR470458-5GEA 13920 CZK
APOSOR470458-5G
Methyl 1-Cbz-3-allylpiperidine-3-carboxylate
Methyl 1-Cbz-3-allylpiperidine-3-carboxylate
Vzorec: C₁₈H₂₃NO₄
MW: 317,38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21641323
CAS číslo: 1363166-19-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR