1-Cbz-3-allylpiperidine-3-carboxylic acid
Dodavatel: Apollo Scientific
APOSOR470459-5GEA
28700
CZK
APOSOR470459-5G
1-Cbz-3-allylpiperidine-3-carboxylic acid
1-Cbz-3-allylpiperidine-3-carboxylic acid
Vzorec:
C₁₇H₂₁NO₄ MW: 303,353 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21641324 CAS číslo: 1363166-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...