Tisk…

2-Ethyl-4-methylbenzoxazole

Dodavatel: Apollo Scientific

APOSOR470460-5GEA 26090 CZK
APOSOR470460-5G
2-Ethyl-4-methylbenzoxazole
2-Ethyl-4-methylbenzoxazole
Vzorec: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21641330
CAS číslo: 1363166-40-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR