Tisk…

2-Ethyl-5-methylbenzoxazole

Dodavatel: Apollo Scientific

APOSOR470466-5GEA 17390 CZK
APOSOR470466-5G
2-Ethyl-5-methylbenzoxazole
2-Ethyl-5-methylbenzoxazole
Vzorec: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19374490
CAS číslo: 20514-29-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR