Tisk…

4-(2-Chlorophenyl)-1,2,3-thiadiazole-5-carbonitrile

Dodavatel: Apollo Scientific

APOSOR470471-1GEA 12770 CZK
APOSOR470471-1G
4-(2-Chlorophenyl)-1,2,3-thiadiazole-5-carbonitrile
4-(2-Chlorophenyl)-1,2,3-thiadiazole-5-carbonitrile
Vzorec: C₉H₄ClN₃S
MW: 221,666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21641340
CAS číslo: 265326-54-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR