Tisk…

Methyl 2-Isopropylbenzoate

Dodavatel: Apollo Scientific

APOSOR470473-1GEA 12770 CZK
APOSOR470473-1G
Methyl 2-Isopropylbenzoate
Methyl 2-Isopropylbenzoate
Vzorec: C₁₁H₁₄O₂
MW: 178,228 g/mol
Bod varu: 249 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD20484275
CAS číslo: 6623-98-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR