Tisk…

4,5-Dihydro-1H-pyrido[2,3-b][1,4]diazepin-2(3H)-one

Dodavatel: Apollo Scientific

APOSOR470479-1GEA 17130 CZK
APOSOR470479-1G
4,5-Dihydro-1H-pyrido[2,3-b][1,4]diazepin-2(3H)-one
4,5-Dihydro-1H-pyrido[2,3-b][1,4]diazepin-2(3H)-one
Vzorec: C₈H₉N₃O
MW: 163,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14705116
CAS číslo: 757966-64-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR