Tisk…

3-Bromo-5-chloro-1-(p-toluenesulphonyl)indole

Dodavatel: Apollo Scientific

APOSOR470495-5GEA 20300 CZK
APOSOR470495-5G
3-Bromo-5-chloro-1-(p-toluenesulphonyl)indole
3-Bromo-5-chloro-1-(p-toluenesulphonyl)indole
Vzorec: C₁₅H₁₁BrClNO₂S
MW: 384,675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22205872
CAS číslo: 887338-49-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR