Tisk…

α-(3-Biphenylyl)benzylamine

Dodavatel: Apollo Scientific

APOSOR470506-1GEA 17130 CZK
APOSOR470506-1G
α-(3-Biphenylyl)benzylamine
α-(3-Biphenylyl)benzylamine
Vzorec: C₁₉H₁₇N
MW: 259,345 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22574976
CAS číslo: 1138152-53-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR