Tisk…

α-(4-Benzyloxyphenyl)benzylamine

Dodavatel: Apollo Scientific

APOSOR470515-5GEA 22920 CZK
APOSOR470515-5G
α-(4-Benzyloxyphenyl)benzylamine
α-(4-Benzyloxyphenyl)benzylamine
Vzorec: C₂₀H₁₉NO
MW: 289,371 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22375699
CAS číslo: 748769-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR