Tisk…

1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylic acid

Dodavatel: Apollo Scientific

APOSOR470521-5GEA 28700 CZK
APOSOR470521-5G
1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylic acid
1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylic acid
Vzorec: C₁₇H₂₂O₃
MW: 274,355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22423119
CAS číslo: 1385694-58-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR