Methyl 1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Dodavatel: Apollo Scientific
APOSOR470523-5GEA
28700
CZK
APOSOR470523-5G
Methyl 1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Vzorec:
C₁₄H₁₄Cl₂O₃ MW: 301,165 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22574979 CAS číslo: 1408058-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...