Tisk…

Methyl 1-(2-Nitrophenyl)-4-oxocyclohexanecarboxylate

Dodavatel: Apollo Scientific

APOSOR470531-5GEA 28700 CZK
APOSOR470531-5G
Methyl 1-(2-Nitrophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Nitrophenyl)-4-oxocyclohexanecarboxylate
Vzorec: C₁₄H₁₅NO₅
MW: 277,273 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22423137
CAS číslo: 1385694-64-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR