1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylic acid
Dodavatel: Apollo Scientific
APOSOR470536-5GEA
31910
CZK
APOSOR470536-5G
1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylic acid
1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylic acid
Vzorec:
C₁₄H₁₅NO₅ MW: 277,273 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22423143 CAS číslo: 1385694-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...