Tisk…

1-(3-Bromophenyl)-4-oxocyclohexanecarboxylic acid

Dodavatel: Apollo Scientific

APOSOR470538-5GEA 28700 CZK
APOSOR470538-5G
1-(3-Bromophenyl)-4-oxocyclohexanecarboxylic acid
1-(3-Bromophenyl)-4-oxocyclohexanecarboxylic acid
Vzorec: C₁₃H₁₃BrO₃
MW: 297,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22423145
CAS číslo: 1385694-53-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR