(3-Pyridyl)(3-quinolyl)methanol
Dodavatel: Apollo Scientific
APOSOR470549-1GEA
14220
CZK
APOSOR470549-1G
(3-Pyridyl)(3-quinolyl)methanol
(3-Pyridyl)(3-quinolyl)methanol
Vzorec:
C₁₅H₁₂N₂O MW: 236,269 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12833812 CAS číslo: 1183608-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...