2-(4-Benzyl-2-morpholinyl)ethanamine
Dodavatel: Apollo Scientific
APOSOR470551-1GEA
26090
CZK
APOSOR470551-1G
2-(4-Benzyl-2-morpholinyl)ethanamine
2-(4-Benzyl-2-morpholinyl)ethanamine
Vzorec:
C₁₃H₂₀N₂O MW: 220,311 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11108389 CAS číslo: 112914-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...