3-Amino-2-indenecarbonitrile
Dodavatel: Apollo Scientific
APOSOR470560-1GEA
12770
CZK
APOSOR470560-1G
3-Amino-2-indenecarbonitrile
3-Amino-2-indenecarbonitrile
Vzorec:
C₁₀H₈N₂ MW: 156,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01104042 CAS číslo: 195067-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...