Tisk…

Methyl 5-Ethoxy-2-thiophenecarboxylate

Dodavatel: Apollo Scientific

APOSOR470579-1GEA 17130 CZK
APOSOR470579-1G
Methyl 5-Ethoxy-2-thiophenecarboxylate
Methyl 5-Ethoxy-2-thiophenecarboxylate
Vzorec: C₈H₁₀O₃S
MW: 186,228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22682792
CAS číslo: 1418117-83-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR