Tisk…

2-(Hydroxymethyl)-N-methylbenzamide

Dodavatel: Apollo Scientific

APOSOR470601-5GEA 20300 CZK
APOSOR470601-5G
2-(Hydroxymethyl)-N-methylbenzamide
2-(Hydroxymethyl)-N-methylbenzamide
Vzorec: C₉H₁₁NO₂
MW: 165,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00157782
CAS číslo: 39976-03-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR