3-(4-Pyridyl)propanamide
Dodavatel: Apollo Scientific
APOSOR470612-5GEA
28400
CZK
APOSOR470612-5G
3-(4-Pyridyl)propanamide
3-(4-Pyridyl)propanamide
Vzorec:
C₈H₁₀N₂O MW: 150,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00956248 CAS číslo: 84200-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...