Tisk…

2-Amino-4-chloro-6-methoxyquinazoline

Dodavatel: Apollo Scientific

APOSOR470648-1GEA 20300 CZK
APOSOR470648-1G
2-Amino-4-chloro-6-methoxyquinazoline
2-Amino-4-chloro-6-methoxyquinazoline
Vzorec: C₉H₈ClN₃O
MW: 209,632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18378817
CAS číslo: 1379296-48-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR