Tisk…

Methyl (S)-2-(Boc-amino)-5-iodopentanoate

Dodavatel: Apollo Scientific

APOSOR470701-1GEA 31910 CZK
APOSOR470701-1G
Methyl (S)-2-(Boc-amino)-5-iodopentanoate
Methyl (S)-2-(Boc-amino)-5-iodopentanoate
Vzorec: C₁₁H₂₀INO₄
MW: 357,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25541687
CAS číslo: 162007-08-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR