Tisk…

4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester

Dodavatel: Apollo Scientific

APOSOR470702-5GEA 17130 CZK
APOSOR470702-5G
4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester
4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester
Vzorec: C₁₆H₂₁BN₂O₂
MW: 284,161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18735902
CAS číslo: 1351478-35-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR